Learn More
Apexbio Technology LLC CNQX disodium salt 479347-85-8 10mg
SDP
Catalog No. 5000091818
Encompass_Preferred
CNQX disodium salt (CAS 479347-85-8) is a competitive antagonist of non-NMDA glutamate receptors specifically targeting AMPA and kainate subtypes with IC50 values of 0 3 M and 1 5 M respectively By inhibiting these receptors CNQX blocks fast excitatory synaptic transmission in the central nervous system In thalamic reticular neurons CNQX induces sustained depolarization while showing minimal effect on ventrobasal neurons In vivo administration of CNQX in rats reduces capsaicin-induced c-fos expression in the trigeminal nucleus caudalis without influencing other reticular regions CNQX is widely used to delineate non-NMDA receptor functions in neurophysiological and behavioral studies
Catalog No. 50-000-91818 Supplier Apexbio Technology LLC Supplier No. B656710
CNQX disodium salt (CAS 479347-85-8) is a competitive antagonist of non-NMDA glutamate receptors specifically targeting AMPA and kainate subtypes with IC50 values of 0 3 M and 1 5 M respectively By inhibiting these receptors CNQX blocks fast excitatory synaptic transmission in the central nervous system In thalamic reticular neurons CNQX induces sustained depolarization while showing minimal effect on ventrobasal neurons In vivo administration of CNQX in rats reduces capsaicin-induced c-fos expression in the trigeminal nucleus caudalis without influencing other reticular regions CNQX is widely used to delineate non-NMDA receptor functions in neurophysiological and behavioral studies