Learn More
Abcam CNQ x disodium salt (mM/ml), AMPA / kainate antagonist (water soluble), 20MM_ML
Catalog No. 502705673
Encompass_Preferred
MW 276.12 Da, Purity >99%. Water soluble, potent, competitive AMPA / kainate receptor antagonist. Also antagonist at NMDA receptor glycine site.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5673 Supplier Abcam Supplier No. AB14448820MM_ML
Add to Cart
Add to Cart
MW 276.12 Da, Purity >99%. Water soluble, potent, competitive AMPA / kainate receptor antagonist. Also antagonist at NMDA receptor glycine site.
The product is subject to the following: Abcam Restricted Use Statement