Learn More
Selleck Chemical LLC CNQX-E0484-5MG
SDP
Catalog No. 503136965
Encompass_Preferred
CNQX (6-cyano-7-nitroquinoxaline-2 3-dione) is an AMPA and kainate receptor antagonist with IC50 values are 0 3 M and 1 5 M for AMPA and kainate receptors respectively CNQX is also an antagonist at the glycine modulatory site on the NMDA receptor complex (IC50 25 M) CNQX shows neuroprotective effects in models of ischemia and inhibits seizure-like activity in hippocampal neurons
Catalog No. 50-313-6965 Supplier Selleck Chemical LLC Supplier No. E04845MG
CNQX (6-cyano-7-nitroquinoxaline-2 3-dione) is an AMPA and kainate receptor antagonist with IC50 values are 0 3 M and 1 5 M for AMPA and kainate receptors respectively CNQX is also an antagonist at the glycine modulatory site on the NMDA receptor complex (IC50 25 M) CNQX shows neuroprotective effects in models of ischemia and inhibits seizure-like activity in hippocampal neurons