Learn More
Aviva Systems Biology Corp CNR1 Antibody - N-terminal region (ARP59994_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59994_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CNR1. Alternate names: CANN6, CB-R, CB1, CB1A, CB1K5, CB1R, CNRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-910