Learn More
Aviva Systems Biology Corp Cnr1 Antibody - C-terminal region (ARP59995_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59995_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cnr1. It was validated on Western Blot. . Alternate names: CB1, CB-R, CB1RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-911