Learn More
Cayman Chemical CnrtInIbhydrochlorIde 10mg
SDP
Catalog No. 502954324
Encompass_Preferred
An irreversible quinazoline-based HER family tyrosine kinase inhibitor with IC50 values of 0.8, 19, and 7 nM for blocking in vitro activity of EGFR, HER2, and HER4, respectively; inhibits the proliferation of various tumor cells
Catalog No. 50-295-4324 Supplier Cayman Chemical Supplier No. 1207610 MG
An irreversible quinazoline-based HER family tyrosine kinase inhibitor with IC50 values of 0.8, 19, and 7 nM for blocking in vitro activity of EGFR, HER2, and HER4, respectively; inhibits the proliferation of various tumor cells