Learn More
Cayman Chemical Canertinib (hydrochloride), Purity: ≥98%, CAS: 289499-45-2, 5 mg
SDP
Catalog No. 502954184
Encompass_Preferred
An irreversible quinazoline-based HER family tyrosine kinase inhibitor with IC50 values of 0.8, 19, and 7 nM for blocking in vitro activity of EGFR, HER2, and HER4, respectively; inhibits the proliferation of various tumor cells
Catalog No. 50-295-4184 Supplier Cayman Chemical Supplier No. 120765 MG
An irreversible quinazoline-based HER family tyrosine kinase inhibitor with IC50 values of 0.8, 19, and 7 nM for blocking in vitro activity of EGFR, HER2, and HER4, respectively; inhibits the proliferation of various tumor cells