Learn More
TARGETMOL CHEMICALS INC CNS-5161 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581642
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CNS-5161 hydrochloride (CNS 5161A) is a new antagonist of NMDA ion-channel. It interacts with the NMDA receptor/ion channel site to produce a noncompetitive blockade of the actions of glutamate. purity: 99%
Catalog No. 50-258-1642 Supplier TARGETMOL CHEMICALS INC Supplier No. T108525MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CNS-5161 hydrochloride (CNS 5161A) is a new antagonist of NMDA ion-channel. It interacts with the NMDA receptor/ion channel site to produce a noncompetitive blockade of the actions of glutamate. purity: 99%