Learn More
Zen Bio Inc CnT-Prime 2D Diff. (CnT-PR-D)
SDP
Catalog No. NC1085756
Encompass
Media designed for proliferation and longevity contain a range of factors that are known to retard the differentiation process.The CnT-PR-D medium is formulated with a totally different balance of growth factors to encourage cells to differentiate quickly and efficiently in 2D culture.
Catalog No. NC1085756 Supplier Zen Bio Inc Supplier No. CNTPRD
Media designed for proliferation and longevity contain a range of factors that are known to retard the differentiation process.The CnT-PR-D medium is formulated with a totally different balance of growth factors to encourage cells to differentiate quickly and efficiently in 2D culture.