Learn More
Zen Bio Inc CnT-Prime Fibroblast Medium (Low serum) | 500ml
SDP
Catalog No. NC1778321
Encompass
CnT-Prime Fibroblast is a 1% serum medium that delivers improved isolation efficiency and proliferation rate of human and animal fibroblasts in comparison with traditional 10% serum media. It was developed using human tissue, but may also be used with other species 
Catalog No. NC1778321 Supplier Zen Bio Inc Supplier No. CNTPRF
CnT-Prime Fibroblast is a 1% serum medium that delivers improved isolation efficiency and proliferation rate of human and animal fibroblasts in comparison with traditional 10% serum media. It was developed using human tissue, but may also be used with other species 