Learn More
Medchemexpress LLC HY-112843 10mg Medchemexpress, CNT2 inhibitor-1 CAS:880155-70-4 Purity:>98%
SDP
Catalog No. 501969189
Encompass_Preferred
Medchemexpress, HY-112843 10mg CNT2 inhibitor-1 CAS:880155-70-4 CNT2 inhibitor-1 is a potent concentrative nucleoside transporter 2 Inhibitor (CNT2), with an IC50 of 640 nM for hCNT2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9189 Supplier Medchemexpress LLC Supplier No. HY11284310MG
Medchemexpress, HY-112843 10mg CNT2 inhibitor-1 CAS:880155-70-4 CNT2 inhibitor-1 is a potent concentrative nucleoside transporter 2 Inhibitor (CNT2), with an IC50 of 640 nM for hCNT2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.