Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HY-17420A 100mg Medchemexpress, Cyclophosphamide (hydrate) CAS:6055-19-2 Purity: 99.97%.
SDP

Catalog No. 501969987
Encompass_Preferred

HY-17420A 100mg Medchemexpress, Cyclophosphamide (hydrate) CAS:6055-19-2 Purity: 99.97%.

Catalog No. 50-196-9987 Supplier Medchemexpress LLC Supplier No. HY17420A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HY-17420A 100mg Medchemexpress, Cyclophosphamide (hydrate) CAS:6055-19-2 Purity: 99.97%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.