Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CNTF Human HEK293 100ug
SDP

Catalog No. 5000195435
Encompass_Preferred

Recombinant human ciliary neurotrophic factor (CNTF) expressed in HEK293 cells and supplied lyophilized as a tag-free protein corresponding to residues M1-M200 (UniProt P26441). Provided for in vitro research with high purity and characterized bioactivity (ED50 0.089-0.324 μg/mL).

  • Tag-free, full-length sequence corresponding to residues M1-M200.
  • Expressed in HEK293 mammalian cells for native folding and post-translational modifications.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Formulated in 20 mM PB, 150 mM NaCl, pH 7.4.
  • Bioactivity validated with ED50 of 0.089-0.324 μg/mL in a TF-1 cell proliferation assay.
  • Recommended storage at -20°C; aliquot and avoid repeated freeze-thaw.

Catalog No. 50-001-95435 Supplier Medchemexpress LLC Supplier No. HYP7145100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ciliary neurotrophic factor (CNTF) expressed in HEK293 cells and supplied lyophilized as a tag-free protein corresponding to residues M1-M200 (UniProt P26441). Provided for in vitro research with high purity and characterized bioactivity (ED50 0.089-0.324 μg/mL).

  • Tag-free, full-length sequence corresponding to residues M1-M200.
  • Expressed in HEK293 mammalian cells for native folding and post-translational modifications.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Formulated in 20 mM PB, 150 mM NaCl, pH 7.4.
  • Bioactivity validated with ED50 of 0.089-0.324 μg/mL in a TF-1 cell proliferation assay.
  • Recommended storage at -20°C; aliquot and avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.