Learn More
Medchemexpress LLC HY-P7148 10ug Medchemexpress, CNTF, Rat CAS: Purity:>98%
SDP
Catalog No. 502083114
Encompass_Preferred
Medchemexpress, HY-P7148 10ug CNTF, Rat CAS: Recombinant Rat Ciliary Neurotrophic Factor, a pluripotent neurotrophic factor, could protect retinal cone and rod photoreceptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3114 Supplier Medchemexpress LLC Supplier No. HYP714810UG
Medchemexpress, HY-P7148 10ug CNTF, Rat CAS: Recombinant Rat Ciliary Neurotrophic Factor, a pluripotent neurotrophic factor, could protect retinal cone and rod photoreceptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.