Learn More
ACROBiosystems Recombinant Protein;Human CNTF R alpha Protein;HEK293;100UG;CNR,CNTFR,CNTF R alpha,CNTFR-alpha
Catalog No. 501658484
Encompass_Preferred
Recombinant Protein;Human CNTF R alpha Protein;100UG;Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Pro 346 (Accession # AAH01492).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8484 Supplier ACROBiosystems Supplier No. CNRH5225100UG
Recombinant Protein;Human CNTF R alpha Protein;100UG;Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Pro 346 (Accession # AAH01492).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.