Learn More
Cayman Chemical Cantharidic Acid (sodium salt), Purity: ≥95%, CAS: 1465-77-6, 50 mg
SDP
Catalog No. 502983616
Encompass_Preferred
A PP1 and PP2A inhibitor with IC50 values of 0.6 and 0.05 µM, respectively
Catalog No. 50-298-3616 Supplier Cayman Chemical Supplier No. 1907350 MG
A PP1 and PP2A inhibitor with IC50 values of 0.6 and 0.05 µM, respectively