Learn More
Cayman Chemical Cantharidin, Purity: ≥98%, CAS: 56-25-7, 100 mg
SDP
Catalog No. 502972316
Encompass_Preferred
Inhibits PP1 (IC50 = 1.7 μM) and PP2A (IC50 = 0.2 μM) while only weakly inhibiting the activity of PP2B (IC50 = 1 mM); stimulates cell cycle progression and induces premature mitosis and has been shown to be active in various cancer cell lines
Catalog No. 50-297-2316 Supplier Cayman Chemical Supplier No. 14589100 MG
Inhibits PP1 (IC50 = 1.7 μM) and PP2A (IC50 = 0.2 μM) while only weakly inhibiting the activity of PP2B (IC50 = 1 mM); stimulates cell cycle progression and induces premature mitosis and has been shown to be active in various cancer cell lines