Learn More
Grainger Cntinmnt Try Black/Black For 24in Shf
Catalog No. 502753082
Encompass_Preferred
The Grainger Approved Containment Tray is a black plastic spill containment tray designed to help contain leaks and drips from stored items. It measures 14" x 20" x 2" and has a load capacity of 75 lbs. The tray features a ribbed bottom to help direct spills to the lowest point and has 3" high walls to help contain leaks. Made from durable polyethylene, this spill containment tray is chemical and corrosion resistant. With its large size, it can hold multiple containers, car parts, or other items that may leak while in storage. This black tray allows for easy visual inspection to identify leaks.
Catalog No. 50-275-3082 Supplier Grainger Supplier No. 14U098
The Grainger Approved Containment Tray is a black plastic spill containment tray designed to help contain leaks and drips from stored items. It measures 14" x 20" x 2" and has a load capacity of 75 lbs. The tray features a ribbed bottom to help direct spills to the lowest point and has 3" high walls to help contain leaks. Made from durable polyethylene, this spill containment tray is chemical and corrosion resistant. With its large size, it can hold multiple containers, car parts, or other items that may leak while in storage. This black tray allows for easy visual inspection to identify leaks.