Learn More
Grainger Cntinmnt Try Black/Blue For 24in Shf
Catalog No. 502751832
Encompass_Preferred
The Tripp Lite 14U 4-Post Open Frame Rack offers a sturdy, organized space for IT equipment in server rooms. It features a 4-post design with adjustable rails for 19" rack equipment up to 35.4" deep. Ventilation slots on the top and bottom promote air circulation. Levelers and casters make it stationary or portable. Dimensions are 24" W x 22" D x 28" H, with a 200 lbs weight capacity per rail pair. Ideal for network switches, patch panels, and servers, the open frame maximizes space efficiency.
Catalog No. 50-275-1832 Supplier Grainger Supplier No. 14U101
The Tripp Lite 14U 4-Post Open Frame Rack offers a sturdy, organized space for IT equipment in server rooms. It features a 4-post design with adjustable rails for 19" rack equipment up to 35.4" deep. Ventilation slots on the top and bottom promote air circulation. Levelers and casters make it stationary or portable. Dimensions are 24" W x 22" D x 28" H, with a 200 lbs weight capacity per rail pair. Ideal for network switches, patch panels, and servers, the open frame maximizes space efficiency.