Learn More
Grainger Cntinmnt Try Black/Red For 24in Shf
Catalog No. 502752574
Encompass_Preferred
The Tripp Lite 14U099 is a 4-post open frame rack that stands 79 inches tall and has an overall width of 23.5 inches. It is constructed from steel with a black powder coat finish and has a weight capacity of 1,000 lbs. The rack has adjustable mounting rails that accommodate 19-inch rack equipment up to 36 inches deep. It also features heavy duty casters that allow it to be easily maneuvered. The open frame design allows efficient airflow and cable management. Overall, the 14U099 provides a sturdy, customizable rack solution for mounting networking and server equipment.
Catalog No. 50-275-2574 Supplier Grainger Supplier No. 14U099
The Tripp Lite 14U099 is a 4-post open frame rack that stands 79 inches tall and has an overall width of 23.5 inches. It is constructed from steel with a black powder coat finish and has a weight capacity of 1,000 lbs. The rack has adjustable mounting rails that accommodate 19-inch rack equipment up to 36 inches deep. It also features heavy duty casters that allow it to be easily maneuvered. The open frame design allows efficient airflow and cable management. Overall, the 14U099 provides a sturdy, customizable rack solution for mounting networking and server equipment.