Learn More
Cayman Chemical Cntla astca Kampfrl 100mg
SDP
Catalog No. 502951723
Encompass_Preferred
An analog of PGF2a containing a 17-phenoxy group on the lower side chain and an isopropyl ester at the C-1 position
Catalog No. 50-295-1723 Supplier Cayman Chemical Supplier No. 11852100 MG
An analog of PGF2a containing a 17-phenoxy group on the lower side chain and an isopropyl ester at the C-1 position