Learn More
Cayman Chemical Kaempferol, Purity: ≥98%, CAS: 520-18-3, 50 mg
SDP
Catalog No. 502951824
Encompass_Preferred
A dietary flavonoid which has diverse physiological activities through both direct and indirect effects; directly binds ERα and ERβ, acting as an inverse agonist or agonist; acts as an antioxidant; suppresses signaling through certain receptor tyrosine kinases, including EGFR and HGF
Catalog No. 50-295-1824 Supplier Cayman Chemical Supplier No. 1185250 MG
A dietary flavonoid which has diverse physiological activities through both direct and indirect effects; directly binds ERα and ERβ, acting as an inverse agonist or agonist; acts as an antioxidant; suppresses signaling through certain receptor tyrosine kinases, including EGFR and HGF