Learn More
Aviva Systems Biology Corp CNTN1 Antibody - middle region (ARP59427_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59427_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNTN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CNTN1. Alternate names: F3, GP135ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-692