Learn More
Aviva Systems Biology Corp CNTNAP1 Antibody - N-terminal region (ARP45482_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45482_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNTNAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CNTNAP1. Alternate names: CASPR, CNTNAP, NRXN4, P190ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59039