Learn More
Aviva Systems Biology Corp CNTNAP3 Antibody - middle region (ARP50201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CNTNAP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CNTNAP3. Alternate names: CASPR3, CNTNAP3A, RP11-138L21.1, RP11-290L7.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60282