Learn More
Med Vet International Jorvet 18mL Specimen Container with Spoon, J1069, 50 Pack
Catalog No. 5000495635
Encompass_Preferred
A proper fecal exam is an essential part of any wellness program. However, getting proper owner compliance is a challenge. The JorVet fecal specimen containers are an effective way to help assist the pet owner in getting a quality stool sample. Clear, plastic vial to view sample Snap-on lid-easy for clients i.e. geriatric to close container Flat bottomed Spoon attached to cap, retrieve stool sample hygienically Patient stick-on label identifies proper client/patient Inexpensive. Great for dispensing to client Note: Not designed for use as a floatation chamber
Catalog No. 50-004-95635 Supplier Med Vet International Supplier No. JOR1069
A proper fecal exam is an essential part of any wellness program. However, getting proper owner compliance is a challenge. The JorVet fecal specimen containers are an effective way to help assist the pet owner in getting a quality stool sample. Clear, plastic vial to view sample Snap-on lid-easy for clients i.e. geriatric to close container Flat bottomed Spoon attached to cap, retrieve stool sample hygienically Patient stick-on label identifies proper client/patient Inexpensive. Great for dispensing to client Note: Not designed for use as a floatation chamber