Learn More
Med Vet International Twirl'em Pack, 3" x 7", J0557, 100-pack
Catalog No. 5000495367
Encompass_Preferred
The affordable sterile container. Ideal for transport of solids, semi-solids, or liquids. Similar to Whirl paks.Heavy-gauge transparent polyethyleneDouble bottom seamCan write on printed surface3" x 7" (76mm x 127mm)4 oz. (120ml) capacity100/case
Catalog No. 50-004-95367 Supplier Med Vet International Supplier No. JOR557
The affordable sterile container. Ideal for transport of solids, semi-solids, or liquids. Similar to Whirl paks.Heavy-gauge transparent polyethyleneDouble bottom seamCan write on printed surface3" x 7" (76mm x 127mm)4 oz. (120ml) capacity100/case