Learn More
Instech Counter-balanced lever arm for mice, 3.5in (9cm)
SDP
Catalog No. 5081220
Encompass
Multi-Axis Counter-Balanced Lever Arms
Designed to minimize stress on microdialysis probes, the Instech counter-balanced lever arms move vertically and horizontally with the animal to prevent slack in the tether. While most other lever arms use a mass as the counterbalance, these use adjustable springs to make them lighter and more responsive.
The mounting plate can be adjusted for mounting to the side or top of many types of cages. The MCLA is a 6-inch (15cm) arm that is typically used with rats. The SMCLA is a 3 1⁄2-inch (9cm) arm that is practically a requirement for any study involving tethered mice. Versions of these balance arms are also available for metabolism cages, operant chambers for IV self-administration and Instech automated blood samplers.
Features
  • Horizontal and vertical movement
  • Lightweight
  • Top or side mounting
Catalog No. 50-812-20 Supplier Instech Supplier No. SMCLA
Multi-Axis Counter-Balanced Lever Arms
Designed to minimize stress on microdialysis probes, the Instech counter-balanced lever arms move vertically and horizontally with the animal to prevent slack in the tether. While most other lever arms use a mass as the counterbalance, these use adjustable springs to make them lighter and more responsive.
The mounting plate can be adjusted for mounting to the side or top of many types of cages. The MCLA is a 6-inch (15cm) arm that is typically used with rats. The SMCLA is a 3 1⁄2-inch (9cm) arm that is practically a requirement for any study involving tethered mice. Versions of these balance arms are also available for metabolism cages, operant chambers for IV self-administration and Instech automated blood samplers.
Features
  • Horizontal and vertical movement
  • Lightweight
  • Top or side mounting