Learn More
Med Vet International Centrifuge (4000 RPM) including Fixed angle rotor 10 ml x 24
Catalog No. 503046619
Encompass_Preferred
This all purpose low speed centrifuge is quiet, compact, and convenient which makes it ideal for any lab. The auto-balancing system extends the centrifuge life and reduces noise and vibration. With a fixed angle rotor option this versatile centrifuge gives you quality and affordability.LED ScreenAuto BalancingBrushless MotorLow MaintenanceProgrammableAutomatic RCF DisplayComes with balance control, lid lock, and temperature/speed protection devicesStainless Steel Chamber
Catalog No. 50-304-6619 Supplier Med Vet International Supplier No. XC40A1
This all purpose low speed centrifuge is quiet, compact, and convenient which makes it ideal for any lab. The auto-balancing system extends the centrifuge life and reduces noise and vibration. With a fixed angle rotor option this versatile centrifuge gives you quality and affordability.LED ScreenAuto BalancingBrushless MotorLow MaintenanceProgrammableAutomatic RCF DisplayComes with balance control, lid lock, and temperature/speed protection devicesStainless Steel Chamber