Learn More
Med Vet International Centrifuge (4000 RPM) including Swing rotor 50 ml x 8
Catalog No. 503045664
Encompass_Preferred
This all purpose low speed centrifuge is quiet, compact, and convenient which makes it ideal for any lab. With swing bucket rotors that can accommodate up to 50mL tubes, this versatile centrifuge can be used in applications from clinical to biochemical for qualitative analysis of serum, plasma, blood, and urine.LED ScreenBrushless MotorLow MaintenanceProgrammableAutomatic RCF DisplayComes with balance control, lid lock, and temperature/speed protection devices
Catalog No. 50-304-5664 Supplier Med Vet International Supplier No. XC50S3
This all purpose low speed centrifuge is quiet, compact, and convenient which makes it ideal for any lab. With swing bucket rotors that can accommodate up to 50mL tubes, this versatile centrifuge can be used in applications from clinical to biochemical for qualitative analysis of serum, plasma, blood, and urine.LED ScreenBrushless MotorLow MaintenanceProgrammableAutomatic RCF DisplayComes with balance control, lid lock, and temperature/speed protection devices