Learn More
Beckman Coulter Open-Top Thinwall Polypropylene Tubes, 16 x 95 mm, 15 mL, 50/pk
Catalog No. NC1088526
Encompass
Open-Top Polypropylene Tube For use in: High Performance and High Capacity Rotors: (Part No. 362396) JS-24.15.
Catalog No. NC1088526 Supplier Beckman Coulter Supplier No. 361707
Open-Top Polypropylene Tube For use in: High Performance and High Capacity Rotors: (Part No. 362396) JS-24.15.