Learn More
Cayman Chemical Cintirorgon, Purity: ≥98%, CAS: 2055536-64-4, 5 mg
SDP
Catalog No. 502939048
Encompass_Preferred
A RORɣ agonist (EC50s = <0.5 µM in a reporter assay and TR-FRET assay)
Catalog No. 50-293-9048 Supplier Cayman Chemical Supplier No. 284255 MG
A RORɣ agonist (EC50s = <0.5 µM in a reporter assay and TR-FRET assay)