Learn More
Grainger Cntrsnk Rtr Bit For No.6 Wd/Scrw 3/8 In
Catalog No. 502766863
Encompass_Preferred
The Grainger Approved Countersink Router Bit for No. 6 Wood Screws is a 1/4" shank high-speed steel countersink bit designed for pre-drilling No. 6 wood screw holes. It features an 82 degree countersink angle and a 1/4" hex shank to fit most routers. The overall length is 2-1/2" and it can be used on wood and wood composites. This is a single flute design that provides smooth, clean cuts.
Catalog No. 50-276-6863 Supplier Grainger Supplier No. 16Y613
The Grainger Approved Countersink Router Bit for No. 6 Wood Screws is a 1/4" shank high-speed steel countersink bit designed for pre-drilling No. 6 wood screw holes. It features an 82 degree countersink angle and a 1/4" hex shank to fit most routers. The overall length is 2-1/2" and it can be used on wood and wood composites. This is a single flute design that provides smooth, clean cuts.