Learn More
Medchemexpress LLC HY-13943 5mg Medchemexpress, CNX-774 CAS:1202759-32-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY139435MG
Encompass_Preferred
Medchemexpress, HY-13943 5mg CNX-774 CAS:1202759-32-7 CNX-774 is an orally active, irreversible and selective BTK inhibitor, with an IC50 of < 1 nM. CNX-774 specifically targets Cysteine 481 of Btk for covalent modification. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1535