Learn More
Medchemexpress LLC Co 101244 hydrochloride | 193356-17-1 | 99.9% | 377.90 | C21H28ClNO3 | 10 MG
SDP
Catalog No. 5000241174
Encompass_Preferred
Co 101244 hydrochloride (PD 174494 hydrochloride) is a selective antagonist of NR2B (GluN2B)-containing NMDA receptors used to study NMDA receptor pharmacology and neuronal signaling in vitro. The hydrochloride salt is characterized by formula C21H28ClNO3, molecular weight 377.90 g/mol, and supplier-reported purity near 99.9%. It is supplied in small-molecule formats for research assays.
  • Selective antagonist of NR2B-containing NMDA receptors.
  • High chemical purity for reproducible experimental results.
  • Hydrochloride salt form for improved solubility in assay solvents.
  • Suitable for in vitro pharmacology and receptor binding assays.
  • Available in mg-scale solid and DMSO solution formats.
  • Molecular weight and formula provided for dosing calculations.
Catalog No. 50-002-41174 Supplier Medchemexpress LLC Supplier No. HY10770610MG
Co 101244 hydrochloride (PD 174494 hydrochloride) is a selective antagonist of NR2B (GluN2B)-containing NMDA receptors used to study NMDA receptor pharmacology and neuronal signaling in vitro. The hydrochloride salt is characterized by formula C21H28ClNO3, molecular weight 377.90 g/mol, and supplier-reported purity near 99.9%. It is supplied in small-molecule formats for research assays.
  • Selective antagonist of NR2B-containing NMDA receptors.
  • High chemical purity for reproducible experimental results.
  • Hydrochloride salt form for improved solubility in assay solvents.
  • Suitable for in vitro pharmacology and receptor binding assays.
  • Available in mg-scale solid and DMSO solution formats.
  • Molecular weight and formula provided for dosing calculations.