Learn More
Medchemexpress LLC Co 101244 hydrochloride | 193356-17-1 | 99.9% | 377.90 g·mol⁻1 | C21H28ClNO3 | 1 ML
SDP
Catalog No. 5000241697
Encompass_Preferred
Co 101244 hydrochloride is a research-grade NMDA receptor antagonist selective for NR2B-containing receptor subtypes. It is used as a pharmacological tool in electrophysiology and biochemical studies to probe NMDA receptor function. The compound is available as a solid and as a 10 mM solution in DMSO for experimental use.
  • Selective antagonist of NR2B-containing NMDA receptors.
  • Provided as 10 mM solution in DMSO (1 mL) for immediate use.
  • High reported purity suitable for research applications.
  • Light yellow solid form for storage and handling.
  • Useful in preclinical electrophysiology and pharmacology studies.
Catalog No. 50-002-41697 Supplier Medchemexpress LLC Supplier No. HY10770610MM 1ML
Co 101244 hydrochloride is a research-grade NMDA receptor antagonist selective for NR2B-containing receptor subtypes. It is used as a pharmacological tool in electrophysiology and biochemical studies to probe NMDA receptor function. The compound is available as a solid and as a 10 mM solution in DMSO for experimental use.
  • Selective antagonist of NR2B-containing NMDA receptors.
  • Provided as 10 mM solution in DMSO (1 mL) for immediate use.
  • High reported purity suitable for research applications.
  • Light yellow solid form for storage and handling.
  • Useful in preclinical electrophysiology and pharmacology studies.