Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Co 101244 (hydrochloride) | 193356-17-1 | 99.9% | 377.90 g/mol | C21H28ClNO3 | 25 MG
SDP

Catalog No. 5000244513
Encompass_Preferred

Co 101244 hydrochloride is the hydrochloride salt of a selective NR2B-containing NMDA receptor antagonist used in pharmacology and neuroscience research. It is supplied as a solid with high purity and is accompanied by supporting documentation for safe handling and experimental use.

  • High purity: 99.9%.
  • Molecular weight 377.90 g/mol.
  • Chemical formula C21H28ClNO3.
  • Appearance solid, light yellow to yellow.
  • Storage recommendations: store sealed and away from moisture; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.
  • Includes datasheet and safety data sheet for handling and hazard information.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-44513 Supplier Medchemexpress LLC Supplier No. HY10770625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Co 101244 hydrochloride is the hydrochloride salt of a selective NR2B-containing NMDA receptor antagonist used in pharmacology and neuroscience research. It is supplied as a solid with high purity and is accompanied by supporting documentation for safe handling and experimental use.

  • High purity: 99.9%.
  • Molecular weight 377.90 g/mol.
  • Chemical formula C21H28ClNO3.
  • Appearance solid, light yellow to yellow.
  • Storage recommendations: store sealed and away from moisture; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.
  • Includes datasheet and safety data sheet for handling and hazard information.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.