Learn More
Medchemexpress LLC Co 101244 hydrochloride (PD 174494) | 193356-17-1 | 99.9% | 377.90 | C21H28ClNO3 | 50 MG
SDP
Catalog No. 5000242041
Encompass_Preferred
Co 101244 hydrochloride is the hydrochloride salt of a small-molecule NR2B-containing NMDA receptor antagonist supplied for research use. It is a light yellow to yellow solid with high purity and characterized molecular properties and storage/solubility data for in vitro and in vivo applications.
  • Selective NR2B NMDA receptor antagonist useful for neuroscience research.
  • High purity suitable for analytical and biological assays.
  • Well-characterized solubility in DMSO for solution preparation.
  • Stable when stored sealed at recommended temperatures.
  • Available in multiple small-scale quantities for laboratory use.
Catalog No. 50-002-42041 Supplier Medchemexpress LLC Supplier No. HY10770650MG
Co 101244 hydrochloride is the hydrochloride salt of a small-molecule NR2B-containing NMDA receptor antagonist supplied for research use. It is a light yellow to yellow solid with high purity and characterized molecular properties and storage/solubility data for in vitro and in vivo applications.
  • Selective NR2B NMDA receptor antagonist useful for neuroscience research.
  • High purity suitable for analytical and biological assays.
  • Well-characterized solubility in DMSO for solution preparation.
  • Stable when stored sealed at recommended temperatures.
  • Available in multiple small-scale quantities for laboratory use.