Learn More
Medchemexpress LLC Co 101244 hydrochloride (PD 174494 hydrochloride) | 193356-17-1 | 99.9% | 377.90 g/mol | C21H28ClNO3 | 100 MG
SDP
Catalog No. 5000242100
Encompass_Preferred
Co 101244 hydrochloride is a research-grade small-molecule antagonist selective for NR2B (GluN2B)-containing NMDA receptors. Supplied as a light yellow solid hydrochloride salt, it is intended for pharmacology and neuroscience research to probe GluN2B-mediated signaling and receptor function. The compound is reported with high purity and is compatible with DMSO-based in vitro studies.
Catalog No. 50-002-42100 Supplier Medchemexpress LLC Supplier No. HY107706100MG
Co 101244 hydrochloride is a research-grade small-molecule antagonist selective for NR2B (GluN2B)-containing NMDA receptors. Supplied as a light yellow solid hydrochloride salt, it is intended for pharmacology and neuroscience research to probe GluN2B-mediated signaling and receptor function. The compound is reported with high purity and is compatible with DMSO-based in vitro studies.