Learn More
Apexbio Technology LLC CO-1686 (AVL-301), 10mg. Cas: 1374640-70-6 MFCD: MFCD26793864. EGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC A332010
Encompass_Preferred
CO-1686 is an irreversible and orally delivered inhibitor of mutant EGFR with IC50 value of < 0.51 nM for recombinant EGFR L858R/T790M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4796