Learn More
Apexbio Technology LLC CO-1686 (AVL-301) 1374640-70-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064391
Encompass_Preferred
CO-1686 (AVL-301 CAS 1374640-70-6) is an orally delivered irreversible small molecule inhibitor targeting mutated epidermal growth factor receptor (EGFR) It functions by forming a covalent bond at cysteine 797 in the ATP-binding domain of EGFR selectively inhibiting mutant variants such as L858R/T790M with reported IC50 values below 0 51 nM CO-1686 preferentially targets mutant EGFR over wild-type demonstrating approximately 22-fold selectivity Preclinical studies using non-small cell lung cancer (NSCLC) cell lines and xenograft models carrying EGFR mutations showed potent anti-proliferative and tumor regression effects Thus CO-1686 represents a valuable research tool in studying mutant EGFR-mediated oncogenesis
Catalog No. 50-000-64391 Supplier Apexbio Technology LLC Supplier No. A33205.1
CO-1686 (AVL-301 CAS 1374640-70-6) is an orally delivered irreversible small molecule inhibitor targeting mutated epidermal growth factor receptor (EGFR) It functions by forming a covalent bond at cysteine 797 in the ATP-binding domain of EGFR selectively inhibiting mutant variants such as L858R/T790M with reported IC50 values below 0 51 nM CO-1686 preferentially targets mutant EGFR over wild-type demonstrating approximately 22-fold selectivity Preclinical studies using non-small cell lung cancer (NSCLC) cell lines and xenograft models carrying EGFR mutations showed potent anti-proliferative and tumor regression effects Thus CO-1686 represents a valuable research tool in studying mutant EGFR-mediated oncogenesis