Learn More
Braintree Scientific Co-axial Breathing Apparatus, includes 2 rat sized masks
SDP
Catalog No. 501954561
Encompass
COAX-4 The COAX-4 (made from durable Delrin) is a co-axial anesthesia breathing apparatus for maintaining the surgical plane that can be used for mouse to rat sized animals. The fresh anesthetic gas enters the side barbed fitting set with 1/16 tubing that limits dead space. The Waste Anesthetic Gas (WAG) is scavenged by the end barbed fitting to the WAG site. The masks (2 included) utilize a replaceable and transparent silicone diaphragm to seal the nose. Accepts standard tubing. Ten feet of silicone tubing is included. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4561 Supplier Braintree Scientific Supplier No. COAX4R
COAX-4 The COAX-4 (made from durable Delrin) is a co-axial anesthesia breathing apparatus for maintaining the surgical plane that can be used for mouse to rat sized animals. The fresh anesthetic gas enters the side barbed fitting set with 1/16 tubing that limits dead space. The Waste Anesthetic Gas (WAG) is scavenged by the end barbed fitting to the WAG site. The masks (2 included) utilize a replaceable and transparent silicone diaphragm to seal the nose. Accepts standard tubing. Ten feet of silicone tubing is included. **Sold for research purposes only and not for human use**