Learn More
Bioworld CO-CHELATED SEPAROPOREAGA
Supplier:  Bioworld 201810472
Encompass
CO-CHELATED SEPAROPORE (AGAROSE): Co-Chelated Separopore is an imminodiacetate (IDA) immobilized metal affinity chromatography (IMAC) matrix that seperates proteins via their differences in affinity to nickel metal ions. Properties: Aff...
Catalog No. 50-197-785