Learn More
CERILLO INC Co-Culture Discovery Bundle (2 trays of 18 duets + 2 refill packs of 36 duets = 108 Duets)
Supplier:  CERILLO INC CCUB108
Encompass_Preferred
Co-Culture Bundle Package is a set of 2x Co-Culture Plate Sysems (36 Duets) and 2x Refill Packs (72 Duets). CoCulture is designed for real-time phenotypic measurements
Catalog No. NC2603352