Learn More
Alkali Scientific ASI™ Co-Culture Dish (JLK), Sterile, TC Treated, 100 x 20mm, 20/Cs
SDP
Catalog No. 503026592
Encompass_Preferred
Co-culture dish (JLK) is designed to study the interactions between two different cell populations in a single dish, provided with an insert mesh to allow bi-directional signal exchange, achieving more in vivo-like environment. ▪ Material (Frame / Mesh): Polystyrene (PS) / Nylon▪ Nominal membrane thickness: 10 ~ 23 μm▪ Pore size: 23 µm▪ Packed in cell culture-treated cell culture dishes 
Catalog No. 50-302-6592 Supplier Alkali Scientific Supplier No. CCD0100
Co-culture dish (JLK) is designed to study the interactions between two different cell populations in a single dish, provided with an insert mesh to allow bi-directional signal exchange, achieving more in vivo-like environment. ▪ Material (Frame / Mesh): Polystyrene (PS) / Nylon▪ Nominal membrane thickness: 10 ~ 23 μm▪ Pore size: 23 µm▪ Packed in cell culture-treated cell culture dishes 