Learn More
Alkali Scientific ASI™ Co-Culture Slide, TC Treated, Sterile, 12/Cs, 2, PS Chamber, DLux Slide, PP Holder
SDP
Catalog No. 503028776
Encompass_Preferred
Co-Culture Slides are an imaging product that is designed to simplify proceed with the Co-culture. Different cell lines are cultivated separately, but the control of the medium volume allows the substances in the medium can be shared with each other. Through Co-culture, you can intended for checking signal exchange, chemotaxis, and molecular transport between cell lines.   Surface materials for Co Culture SlideDLux: surface modified plastic with minimal autofluorescenceFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy ▪ Convenient for microscopic observation▪ Bottom materials: DLux/FLux▪ Easy open flip for chamber & slide disassembly▪ No chemical adhesives used▪ Packing trays can be used as incubation racks in CO2 incubators▪ Domestic sales only▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-8776 Supplier Alkali Scientific Supplier No. TCC2402
Co-Culture Slides are an imaging product that is designed to simplify proceed with the Co-culture. Different cell lines are cultivated separately, but the control of the medium volume allows the substances in the medium can be shared with each other. Through Co-culture, you can intended for checking signal exchange, chemotaxis, and molecular transport between cell lines.   Surface materials for Co Culture SlideDLux: surface modified plastic with minimal autofluorescenceFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy ▪ Convenient for microscopic observation▪ Bottom materials: DLux/FLux▪ Easy open flip for chamber & slide disassembly▪ No chemical adhesives used▪ Packing trays can be used as incubation racks in CO2 incubators▪ Domestic sales only▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free