Learn More
Lascar Electronics Inc Carbon Monoxide Data Logger with USB
SDP
Catalog No. NC1856629
Encompass_Preferred
This standalone data logger measures and stores up to 32,510 Carbon Monoxide (CO) readings over a 0 to 300ppm measurement range and -10 to +40°C (14 to +104°F) operating temperature range.
Catalog No. NC1856629 Supplier Lascar Electronics Inc Supplier No. ELUSBCO300
This standalone data logger measures and stores up to 32,510 Carbon Monoxide (CO) readings over a 0 to 300ppm measurement range and -10 to +40°C (14 to +104°F) operating temperature range.