Learn More
Bioworld Co-IMAC 5 x 3mL Columns, 5 Column
Catalog No. 50197633
Encompass_Preferred
Co-immobilized metal affinity chromatography (IMAC) columns consist of Co-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Co-IMAC Columns separate proteins via their differences in affinity toCo2+ metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged withCo metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 4BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Cu2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.
Catalog No. 50-197-633 Supplier Bioworld Supplier No. 201700131
Co-immobilized metal affinity chromatography (IMAC) columns consist of Co-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Co-IMAC Columns separate proteins via their differences in affinity toCo2+ metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged withCo metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 4BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Cu2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.