Learn More
Grainger Bulk Strap Bungee Hook, Adjustable, Co-Polymer, 3/16 in. and 1/4 in. Compatibility, 10/Pk.
Catalog No. 502753502
Encompass_Preferred
This adjustable bungee hook is designed for light to medium duty tie down applications, providing a versatile solution for cord management and cargo control. Constructed from durable co-polymer, this hook offers a lightweight yet strong design suitable for a variety of industries including construction, marine, and logistics.
  • Compatible with 3/16 in. and 1/4 in. cords
  • Features a super adjustable design for flexible use
  • Constructed from strong and durable co-polymer
  • Lightweight material composition
  • Overall length of 2 5/16 in. and width of 1 1/4 in.
  • Supplied as a 10-pack
Catalog No. 50-275-3502 Supplier Grainger Supplier No. 14A761
This adjustable bungee hook is designed for light to medium duty tie down applications, providing a versatile solution for cord management and cargo control. Constructed from durable co-polymer, this hook offers a lightweight yet strong design suitable for a variety of industries including construction, marine, and logistics.
  • Compatible with 3/16 in. and 1/4 in. cords
  • Features a super adjustable design for flexible use
  • Constructed from strong and durable co-polymer
  • Lightweight material composition
  • Overall length of 2 5/16 in. and width of 1 1/4 in.
  • Supplied as a 10-pack