Learn More
BINDER™ CO2 Gassing Module
SDP
Catalog No. 502875752 Shop All BINDER Inc. Products
Change view
Click to view available options
CO2 Concentration Range:
0 to 1%
0 to 20%
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CO2 Concentration Range
50-287-5752 0 to 1%
50-287-5600 0 to 20%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-287-5752 Supplier BINDER™ Supplier No. 80122140
Add to Cart
Add to Cart
Request a Quote
CO2 gassing module
  • CO2 gassing module with NDIR CO2 sensor and analog output 4-20 mA
  • Choose between 0 to 20% and 0 to 1%
Specifications
CO2 Concentration Range 0 to 1%
For Use With (Equipment) KBF 470/720/1060/1600, KBF PRO 260/470/720/1060/1600
Material Stainless Steel
Type Gassing Module
Gas Type CO2
Includes NDIR CO2 Sensor and Analog Output 4 to 20 mA